siRNA Details
| Gene Symbol: Cep41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ278891.1 | - | 7 | 511 | ||
| AK006059.1 | - | 7 | 499 | |||
| AK132495.1 | - | 7 | 500 | |||
| AK161461.1 | - | 7 | 485 | |||
| AK166974.1 | - | 8 | 628 | |||
| AK170213.1 | - | 7 | 524 | |||
| BC031892.1 | - | 7 | 481 | 5365552 | ||

Cep41
Genomic Map