siRNA Details
| Gene Symbol: Cep41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ278891.1 | - | 11 | 1482 | ||
| AK132495.1 | - | 11 | 1472 | |||
| AK161461.1 | - | 11 | 1456 | |||
| AK166974.1 | - | 12 | 1599 | |||
| BC031892.1 | - | 11 | 1452 | 5365552 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cep41| Gene Symbol: Cep41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AJ278891.1 | - | 11 | 1482 | ||
| AK132495.1 | - | 11 | 1472 | |||
| AK161461.1 | - | 11 | 1456 | |||
| AK166974.1 | - | 12 | 1599 | |||
| BC031892.1 | - | 11 | 1452 | 5365552 | ||