siRNA Details
Gene Symbol: Mmp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032007.3 | NP_114396.3 | 7 | 967 | ||
GenBank mRNA | AJ278461.1 | - | 7 | 936 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mmp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032007.3 | NP_114396.3 | 7 | 967 | ||
GenBank mRNA | AJ278461.1 | - | 7 | 936 |