siRNA Details
| Gene Symbol: Slmap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032008.3 | NP_114397.3 | 14 | 1540 | ||
| GenBank mRNA | AF304451.1 | - | 2 | 88 | ||
| AK044358.1 | - | 2 | 94 | |||
| AK129403.1 | - | 14 | 1540 | |||
| AK141597.1 | - | 15 | 2760 | |||
| AK146685.1 | - | 2 | 54 | |||
| BC021457.1 | - | 4 | 303 | 5064534 | ||

Slmap
Genomic Map