siRNA Details
| Gene Symbol: Kremen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032396.3 | NP_115772.2 | 5 | 641 | ||
| GenBank mRNA | AB059617.1 | - | 5 | 604 | ||
| AK083565.1 | - | 3 | 249 | |||
| AK141321.1 | - | 5 | 653 | |||
| BC049771.1 | - | 5 | 565 | |||
| BC082546.1 | - | 5 | 641 | 6855459 | ||
| BC138464.1 | - | 5 | 649 | 8861484 | ||
| BC138465.1 | - | 5 | 649 | 8861485 | ||

Kremen1
Genomic Map