siRNA Details
| Gene Symbol: Gpr87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF295366.1 | - | 1 | 327 | ||
| AK080394.1 | - | 3 | 665 | |||
| BC125653.1 | - | 1 | 356 | 40130168 | ||
| BC125655.1 | - | 1 | 356 | 40130170 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr87| Gene Symbol: Gpr87 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF295366.1 | - | 1 | 327 | ||
| AK080394.1 | - | 3 | 665 | |||
| BC125653.1 | - | 1 | 356 | 40130168 | ||
| BC125655.1 | - | 1 | 356 | 40130170 | ||