siRNA Details
Gene Symbol: Gpr87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF295366.1 | - | 1 | 484 | ||
AK080394.1 | - | 3 | 822 | |||
BC125653.1 | - | 1 | 513 | 40130168 | ||
BC125655.1 | - | 1 | 513 | 40130170 |
