siRNA Details
Gene Symbol: Sucnr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032400.2 | NP_115776.2 | 2 | 1159 | ||
GenBank mRNA | AF295367.1 | - | 2 | 1175 | ||
AK080866.1 | - | 2 | 1173 | |||
BC096665.1 | - | 2 | 1132 | 1365839 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
