siRNA Details
| Gene Symbol: Lztfl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033322.2 | NP_201579.1 | 10 | 1277 | ||
| GenBank mRNA | AJ297352.1 | - | 10 | 1273 | ||
| AK029671.1 | - | 10 | 1312 | |||
| AK030068.1 | - | 10 | 1289 | |||
| AK041091.1 | - | 10 | 1310 | |||
| AK046515.1 | - | 10 | 1266 | |||
| AK054003.1 | - | 10 | 1283 | |||
| AK081529.1 | - | 8 | 1062 | |||
| AK145263.1 | - | 10 | 1283 | |||
| AK147312.1 | - | 9 | 1221 | |||
| BC026840.1 | - | 10 | 1277 | 4165152 | ||
| BC037045.1 | - | 10 | 1277 | |||
| BC098224.1 | - | 10 | 1282 | 1263280 | ||

Lztfl1
Genomic Map