siRNA Details
| Gene Symbol: Wbscr16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033572.2 | NP_291050.1 | 4, 5 | 702 | ||
| GenBank mRNA | AF410456.1 | - | 4, 5 | 681 | ||
| AK005040.1 | - | 4, 5 | 648 | |||
| AK017733.1 | - | 4, 5 | 701 | |||
| BC024714.1 | - | 4, 5 | 674 | 5134834 | ||
| BC055335.1 | - | 4, 5 | 700 | 6412407 | ||

Wbscr16
Genomic Map