siRNA Details
| Gene Symbol: Pcdhgb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033580.2 | NP_291058.1 | 1 | 1255 | ||
| GenBank mRNA | AY013810.1 | - | 1 | 1177 | ||
| BC139099.1 | - | 1 | 1255 | 8862121 | ||
| BC139100.1 | - | 1 | 1255 | 8862122 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcdhgb8| Gene Symbol: Pcdhgb8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033580.2 | NP_291058.1 | 1 | 1255 | ||
| GenBank mRNA | AY013810.1 | - | 1 | 1177 | ||
| BC139099.1 | - | 1 | 1255 | 8862121 | ||
| BC139100.1 | - | 1 | 1255 | 8862122 | ||