siRNA Details
| Gene Symbol: Rnf111 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033604.2 | NP_291082.1 | 5 | 1402 | ||
| GenBank mRNA | AF330197.1 | - | 5 | 1398 | ||
| AK036351.1 | - | 4 | 1137 | |||
| AK048110.1 | - | 4 | 1168 | |||
| AK137148.1 | - | 3 | 740 | |||
| AK169999.1 | - | 4 | 1512 | |||
| AK171932.1 | - | 4 | 1340 | |||
| BC069835.1 | - | 5 | 1258 | 30094289 | ||

Rnf111
Genomic Map