siRNA Details
| Gene Symbol: Dach2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142570.1 | NP_001136042.1 | 7, 8 | 1474 | ||
| NM_001289733.1 | NP_001276662.1 | 9, 10 | 1191 | |||
| NM_033605.2 | NP_291083.1 | 8, 9 | 1513 | |||
| GenBank mRNA | AF257217.1 | - | 8, 9 | 1512 | ||
| AK038687.1 | - | 3, 4 | 287 | |||
| AK041446.1 | - | 9, 10 | 1180 | |||
| BC059233.1 | - | 7, 8 | 1463 | 6406445 | ||

Dach2
Genomic Map