siRNA Details
| Gene Symbol: Trim8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053100.2 | NP_444330.2 | 6 | 2406 | ||
| GenBank mRNA | AF281047.1 | - | 6 | 1755 | ||
| AK052763.1 | - | 6 | 1141 | |||
| AK079847.1 | - | 4 | 845 | |||
| AK155303.1 | - | 6 | 2353 | |||
| BC060094.1 | - | 6 | 1636 | |||
| BC082298.1 | - | 6 | 2281 | |||
| BC152343.1 | - | 6 | 1742 | 40093266 | ||
| BC152344.1 | - | 6 | 1742 | 40093269 | ||

Trim8
Genomic Map