siRNA Details
| Gene Symbol: Immp2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053122.4 | NP_444352.2 | 1119 | |||
| GenBank mRNA | AK053361.1 | - | 1093 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Immp2l| Gene Symbol: Immp2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053122.4 | NP_444352.2 | 1119 | |||
| GenBank mRNA | AK053361.1 | - | 1093 | |||