siRNA Details
| Gene Symbol: RP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006269.1 | NP_006260.1 | 3 | 823 | ||
| XM_017013721.1 | XP_016869210.1 | 8 | 1986 | |||
| XM_017013722.1 | XP_016869211.1 | 3 | 839 | |||
| GenBank mRNA | AF141021.1 | - | 3 | 823 | ||
| AF143222.1 | - | 3 | 773 | |||
| AF146592.1 | - | 3 | 771 | |||
| AK098176.1 | - | 5 | 607 | |||
| AK302348.1 | - | 2 | 207 | |||
| AK304443.1 | - | 4 | 368 | |||
| AK316511.1 | - | 4 | 802 | |||

RP1
Genomic Map