siRNA Details
| Gene Symbol: Mrpl43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053164.3 | NP_444394.1 | 2 | 230 | ||
| GenBank mRNA | AB049657.1 | - | 2 | 191 | ||
| AK015357.1 | - | 2 | 211 | |||
| AK076204.1 | - | 2 | 189 | |||
| BC027676.1 | - | 2 | 170 | |||
| BC048733.1 | - | 2 | 221 | 6512868 | ||
| BC065812.1 | - | 2 | 177 | |||

Mrpl43
Genomic Map