siRNA Details
| Gene Symbol: Trim16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053169.2 | NP_444399.2 | 2 | 761 | ||
| GenBank mRNA | AF449496.1 | - | 2 | 512 | ||
| AK053139.1 | - | 2 | 575 | |||
| BC052821.1 | - | 3 | 463 | 30057830 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trim16| Gene Symbol: Trim16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053169.2 | NP_444399.2 | 2 | 761 | ||
| GenBank mRNA | AF449496.1 | - | 2 | 512 | ||
| AK053139.1 | - | 2 | 575 | |||
| BC052821.1 | - | 3 | 463 | 30057830 | ||