siRNA Details
| Gene Symbol: Csmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053171.2 | NP_444401.2 | 67 | 10680 | ||
| GenBank mRNA | AK082377.1 | - | 4 | 478 | ||
| AK220429.1 | - | 18 | 2878 | |||
| AY017475.1 | - | 67 | 10577 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Csmd1| Gene Symbol: Csmd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053171.2 | NP_444401.2 | 67 | 10680 | ||
| GenBank mRNA | AK082377.1 | - | 4 | 478 | ||
| AK220429.1 | - | 18 | 2878 | |||
| AY017475.1 | - | 67 | 10577 | |||