siRNA Details
| Gene Symbol: Ddx50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053183.2 | NP_444413.1 | 2 | 316 | ||
| GenBank mRNA | AF334104.1 | - | 2 | 317 | ||
| AK041281.1 | - | 2 | 215 | |||
| AK162423.1 | - | 3 | 434 | |||
| BC049906.1 | - | 2 | 318 | |||
| BC055440.1 | - | 2 | 310 | |||
| BC075714.1 | - | 2 | 268 | |||

Ddx50
Genomic Map