siRNA Details
| Gene Symbol: Ric8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053194.4 | NP_444424.1 | 3 | 345 | ||
| GenBank mRNA | AF288813.1 | - | 3 | 344 | ||
| AK031466.1 | - | 3 | 292 | |||
| AK080192.1 | - | 3 | 288 | |||
| AK155282.1 | - | 3 | 320 | |||
| AK165536.1 | - | 3 | 322 | |||
| AK167746.1 | - | 3 | 348 | |||
| AK169367.1 | - | 3 | 309 | |||
| AK171093.1 | - | 1 | 637 | |||
| AK188871.1 | - | 1 | 16 | |||
| BC022917.1 | - | 3 | 322 | 4460521 | ||
| BY256573.1 | - | 3 | 348 | |||

Ric8
Genomic Map