siRNA Details
| Gene Symbol: MYL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006097.4 | NP_006088.2 | 2 | 217 | ||
| NM_181526.2 | NP_852667.1 | 2 | 217 | |||
| GenBank mRNA | AF176042.1 | - | 1 | 153 | ||
| AF176043.1 | - | 1 | 153 | |||
| AK097235.1 | - | 1 | 3473 | |||
| AK130879.1 | - | 2 | 182 | |||
| AL697699.1 | - | 2 | 217 | |||
| BC002648.2 | - | 2 | 156 | 3608316 | ||
| BQ953325.1 | - | 207 | ||||
| CR456843.1 | - | 1 | 113 | |||
| D82057.1 | - | 2 | 185 | |||
| J02854.1 | - | 1 | 155 | |||

MYL9
Genomic Map