siRNA Details
| Gene Symbol: Msi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054043.3 | NP_473384.1 | 14 | 1331 | ||
| GenBank mRNA | AK007122.1 | - | 4 | 540 | ||
| AK049637.1 | - | 14 | 1277 | |||
| AK049688.1 | - | 14 | 1277 | |||
| AK136156.1 | - | 10 | 1139 | |||
| AK153928.1 | - | 14 | 1204 | |||
| AK169846.1 | - | 14 | 1204 | |||

Msi2
Genomic Map