siRNA Details
| Gene Symbol: Adgra3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054044.2 | NP_473385.2 | 19 | 5309 | ||
| GenBank mRNA | AF378759.1 | - | 19 | 5309 | ||
| AK018412.1 | - | 7 | 1208 | |||
| AK020642.1 | - | 1 | 678 | |||
| CJ243065.1 | - | 1 | 242 | |||
More Information
Set Membership: |
|

Adgra3
Genomic Map