siRNA Details
| Gene Symbol: Zfand2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159908.1 | NP_001153380.1 | 3 | 369 | ||
| NM_133349.3 | NP_579927.1 | 3 | 380 | |||
| GenBank mRNA | AF224494.1 | - | 3 | 266 | ||
| AK146082.1 | - | 3 | 318 | |||
| AK171245.1 | - | 3 | 320 | |||
| BC058780.1 | - | 3 | 306 | 6312202 | ||

Zfand2a
Genomic Map