siRNA Details
| Gene Symbol: Wwtr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168281.1 | NP_001161753.1 | 4 | 943 | ||
| NM_133784.3 | NP_598545.2 | 4 | 1022 | |||
| GenBank mRNA | AJ299430.1 | - | 4 | 957 | ||
| AK009983.1 | - | 4 | 953 | |||
| AK011504.1 | - | 4 | 996 | |||
| AK142299.1 | - | 4 | 944 | |||
| AK142720.1 | - | 4 | 947 | |||
| BC004640.1 | - | 4 | 943 | 3157246 | ||
| BC014727.1 | - | 4 | 974 | 4487316 | ||

Wwtr1
Genomic Map