siRNA Details
| Gene Symbol: R74862 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034864.1 | - | 5 | 1094 | ||
| AK035397.1 | - | 4 | 650 | |||
| AK083267.1 | - | 4 | 699 | |||
| BC005669.1 | - | 4 | 384 | |||
| BC080794.1 | - | 4 | 967 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

R74862| Gene Symbol: R74862 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK034864.1 | - | 5 | 1094 | ||
| AK035397.1 | - | 4 | 650 | |||
| AK083267.1 | - | 4 | 699 | |||
| BC005669.1 | - | 4 | 384 | |||
| BC080794.1 | - | 4 | 967 | |||