siRNA Details
| Gene Symbol: Nol12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133800.3 | NP_598561.2 | 6 | 1297 | ||
| GenBank mRNA | AK036891.1 | - | 6 | 1240 | ||
| AK037035.1 | - | 6 | 1291 | |||
| AK083539.1 | - | 1299 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nol12| Gene Symbol: Nol12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133800.3 | NP_598561.2 | 6 | 1297 | ||
| GenBank mRNA | AK036891.1 | - | 6 | 1240 | ||
| AK037035.1 | - | 6 | 1291 | |||
| AK083539.1 | - | 1299 | ||||