siRNA Details
| Gene Symbol: Lrrc59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133807.1 | NP_598568.1 | 7 | 1565 | ||
| GenBank mRNA | AK053450.1 | - | 7 | 1592 | ||
| AK159374.1 | - | 7 | 1621 | |||
| AK159552.1 | - | 7 | 1615 | |||
| AK166576.1 | - | 7 | 1614 | |||
| AK167604.1 | - | 7 | 1556 | |||
| AK169250.1 | - | 5 | 1177 | |||
| AK169390.1 | - | 7 | 1564 | |||
| BC006877.1 | - | 7 | 1565 | 2647513 | ||
| BC066172.1 | - | 7 | 1559 | 30429192 | ||

Lrrc59
Genomic Map