siRNA Details
| Gene Symbol: Stk17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133810.3 | NP_598571.2 | 5 | 691 | ||
| GenBank mRNA | AK035693.1 | - | 5 | 659 | ||
| AK041139.1 | - | 1 | 26 | |||
| AK076932.1 | - | 5 | 658 | |||
| AK083332.1 | - | 5 | 660 | |||
| AK153108.1 | - | 5 | 635 | |||
| AK165291.1 | - | 5 | 631 | |||
| AY092028.1 | - | 5 | 644 | |||
| BC006579.1 | - | 5 | 646 | 3586767 | ||

Stk17b
Genomic Map