siRNA Details
| Gene Symbol: Stk17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133810.3 | NP_598571.2 | 8 | 2744 | ||
| GenBank mRNA | AY092028.1 | - | 8 | 2697 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Stk17b| Gene Symbol: Stk17b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133810.3 | NP_598571.2 | 8 | 2744 | ||
| GenBank mRNA | AY092028.1 | - | 8 | 2697 | ||