siRNA Details
| Gene Symbol: AI597479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133818.1 | NP_598579.1 | 2 | 206 | ||
| GenBank mRNA | AK081372.1 | - | 2 | 197 | ||
| BC006931.1 | - | 2 | 206 | 3598975 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

AI597479| Gene Symbol: AI597479 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133818.1 | NP_598579.1 | 2 | 206 | ||
| GenBank mRNA | AK081372.1 | - | 2 | 197 | ||
| BC006931.1 | - | 2 | 206 | 3598975 | ||