siRNA Details
| Gene Symbol: Cdc123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133837.4 | NP_598598.1 | 8, 9 | 618 | ||
| GenBank mRNA | AK090122.1 | - | 8, 9 | 619 | ||
| AK141463.1 | - | 8, 9 | 606 | |||
| AK159208.1 | - | 8, 9 | 639 | |||
| BC019753.1 | - | 8, 9 | 593 | 5008314 | ||
| BC022972.1 | - | 8, 9 | 607 | 5368775 | ||
| BC023821.1 | - | 8, 9 | 610 | 5354711 | ||
| BC024787.1 | - | 8, 9 | 605 | 5359215 | ||

Cdc123
Genomic Map