siRNA Details
| Gene Symbol: Usp53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133857.3 | NP_598618.1 | 7 | 1343 | ||
| GenBank mRNA | AK030317.1 | - | 8 | 1427 | ||
| AK036876.1 | - | 7 | 1344 | |||
| AK040332.1 | - | 8 | 1239 | |||
| AK045953.1 | - | 7 | 1406 | |||
| AK140453.1 | - | 7 | 1387 | |||
| BC096383.1 | - | 7 | 1343 | |||
| BC132339.1 | - | 6 | 1076 | 40130616 | ||
| BC145707.1 | - | 6 | 1076 | 40131010 | ||

Usp53
Genomic Map