siRNA Details
| Gene Symbol: Eps8l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133867.2 | NP_598628.1 | 5 | 460 | ||
| GenBank mRNA | AK136584.1 | - | 5 | 456 | ||
| AK167630.1 | - | 5 | 447 | |||
| AY074932.1 | - | 5 | 476 | |||
| BC014734.1 | - | 5 | 448 | 4218079 | ||

Eps8l3
Genomic Map