siRNA Details
| Gene Symbol: Ifi44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133871.2 | NP_598632.2 | 2 | 464 | ||
| GenBank mRNA | AJ299405.1 | - | 1 | 354 | ||
| AK079749.1 | - | 2 | 416 | |||
| AK085407.1 | - | 2 | 453 | |||
| AK085650.1 | - | 2 | 398 | |||
| AK136553.1 | - | 2 | 416 | |||
| AK151346.1 | - | 2 | 416 | |||
| AK171676.1 | - | 2 | 397 | |||
| BB645803.1 | - | 2 | 453 | |||
| BC111882.1 | - | 1 | 354 | 40060245 | ||
| BC112402.1 | - | 1 | 354 | |||

Ifi44
Genomic Map