siRNA Details
| Gene Symbol: Gpn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290742.1 | NP_001277671.1 | 2 | 451 | ||
| NM_133884.2 | NP_598645.2 | 2 | 520 | |||
| GenBank mRNA | BC018407.1 | - | 2 | 513 | 4015340 | |
| CN721622.1 | - | 2 | 122 | |||
| CO798620.2 | - | 2 | 520 | |||

Gpn2
Genomic Map