siRNA Details
| Gene Symbol: Emilin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133918.2 | NP_598679.1 | 2 | 330 | ||
| GenBank mRNA | AK029337.1 | - | 2 | 301 | ||
| AK052368.1 | - | 2 | 411 | |||
| AK088995.1 | - | 2 | 394 | |||
| AK155486.1 | - | 2 | 632 | |||
| BC005481.1 | - | 2 | 330 | 3485057 | ||

Emilin1
Genomic Map