siRNA Details
| Gene Symbol: Krba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133922.3 | NP_598683.3 | 6 | 970 | ||
| GenBank mRNA | AB041543.1 | - | 5 | 800 | ||
| AK040525.1 | - | 6 | 972 | |||
| AK128931.1 | - | 6 | 921 | |||
| AK162934.1 | - | 6 | 678 | |||
| AK173281.1 | - | 6 | 2674 | |||
| BC066816.1 | - | 6 | 890 | 30084911 | ||

Krba1
Genomic Map