siRNA Details
| Gene Symbol: Krba1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133922.3 | NP_598683.3 | 11, 12 | 1884 | ||
| GenBank mRNA | AB041543.1 | - | 9, 10 | 1516 | ||
| AK029235.1 | - | 2, 3 | 330 | |||
| AK040525.1 | - | 11, 12 | 1886 | |||
| AK128931.1 | - | 11, 12 | 1835 | |||
| AK162934.1 | - | 11, 12 | 1592 | |||
| AK173281.1 | - | 11, 12 | 3588 | |||
| BC066816.1 | - | 11, 12 | 1804 | 30084911 | ||

Krba1
Genomic Map