siRNA Details
| Gene Symbol: Vrk3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133945.1 | NP_598706.1 | 15 | 1516 | ||
| GenBank mRNA | AK150435.1 | - | 15 | 1607 | ||
| AK171673.1 | - | 15 | 1521 | |||
| BC010473.1 | - | 14 | 1499 | 3586304 | ||
| BC024782.1 | - | 14 | 1499 | 5123949 | ||
| BC024839.1 | - | 15 | 1516 | 5355521 | ||
| BC027129.1 | - | 13 | 1285 | |||
| BC034196.1 | - | 14 | 1497 | 4985176 | ||
| S71575.1 | - | 7 | 751 | |||

Vrk3
Genomic Map