siRNA Details
| Gene Symbol: Usb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133954.2 | NP_598715.2 | 7 | 1212 | ||
| GenBank mRNA | AK135527.1 | - | 7 | 1205 | ||
| AK160901.1 | - | 7 | 1195 | |||
| AK168683.1 | - | 7 | 1195 | |||
| BC016418.1 | - | 7 | 1156 | 4224590 | ||

Usb1
Genomic Map