siRNA Details
| Gene Symbol: Zdhhc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133967.3 | NP_598728.1 | 8 | 1940 | ||
| GenBank mRNA | AK041905.1 | - | 8 | 1809 | ||
| AK090333.1 | - | 8 | 1786 | |||
| AK157968.1 | - | 8 | 1793 | |||
| AK170639.1 | - | 8 | 1793 | |||
| AK170923.1 | - | 8 | 1765 | |||
| AK172462.1 | - | 8 | 1774 | |||
| BC013467.1 | - | 7 | 1491 | 4218025 | ||
| BC029666.1 | - | 3 | 894 | |||
| BC075666.1 | - | 8 | 1940 | 30605163 | ||

Zdhhc7
Genomic Map