siRNA Details
| Gene Symbol: Pwp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133993.3 | NP_598754.2 | 5 | 597 | ||
| GenBank mRNA | AK009972.1 | - | 5 | 564 | ||
| AK011891.1 | - | 5 | 545 | |||
| AK082158.1 | - | 5 | 539 | |||
| AK149699.1 | - | 5 | 541 | |||
| BC003199.1 | - | 5 | 566 | 3586496 | ||
| BC023137.1 | - | 5 | 547 | 3986908 | ||

Pwp1
Genomic Map