siRNA Details
| Gene Symbol: Psme4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134013.3 | NP_598774.2 | 4, 5 | 752 | ||
| GenBank mRNA | AF296169.1 | - | 4, 5 | 463 | ||
| BC141381.1 | - | 4, 5 | 465 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psme4| Gene Symbol: Psme4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134013.3 | NP_598774.2 | 4, 5 | 752 | ||
| GenBank mRNA | AF296169.1 | - | 4, 5 | 463 | ||
| BC141381.1 | - | 4, 5 | 465 | |||