siRNA Details
| Gene Symbol: Tbc1d10a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134023.1 | NP_598784.1 | 9 | 1605 | ||
| GenBank mRNA | AK140872.1 | - | 9 | 1617 | ||
| AK153677.1 | - | 1393 | ||||
| AK220243.1 | - | 2153 | ||||
| BC018300.1 | - | 9 | 1605 | 4193046 | ||

Tbc1d10a
Genomic Map