siRNA Details
| Gene Symbol: Ssx2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253768.1 | NP_001240697.1 | 5 | 954 | ||
| NM_001253769.1 | NP_001240698.1 | 4 | 850 | |||
| NM_001253770.1 | NP_001240699.1 | 4 | 699 | |||
| NM_138744.3 | NP_620083.1 | 4 | 699 | |||
| GenBank mRNA | AF532969.1 | - | 3 | 494 | ||
| AK031356.1 | - | 5 | 678 | |||
| AK043865.1 | - | 4 | 803 | |||
| AK049080.1 | - | 4 | 851 | |||
| AK077112.1 | - | 5 | 739 | |||
| AK173064.1 | - | 4 | 686 | |||
| BC021749.1 | - | 4 | 654 | 4165430 | ||
| BC031527.1 | - | 4 | 662 | 4008624 | ||

Ssx2ip
Genomic Map