siRNA Details
| Gene Symbol: Ssx2ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253768.1 | NP_001240697.1 | 7 | 1113 | ||
| NM_001253769.1 | NP_001240698.1 | 6 | 1009 | |||
| NM_001253770.1 | NP_001240699.1 | 6 | 858 | |||
| NM_138744.3 | NP_620083.1 | 6 | 858 | |||
| GenBank mRNA | AF532969.1 | - | 5 | 653 | ||
| AK031356.1 | - | 7 | 837 | |||
| AK043865.1 | - | 6 | 962 | |||
| AK049080.1 | - | 6 | 1010 | |||
| AK077112.1 | - | 7 | 898 | |||
| AK173064.1 | - | 6 | 845 | |||
| BC021749.1 | - | 6 | 813 | 4165430 | ||
| BC031527.1 | - | 6 | 821 | 4008624 | ||

Ssx2ip
Genomic Map