siRNA Details
| Gene Symbol: 2210407C18Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144544.2 | NP_653127.2 | 3 | 185 | ||
| GenBank mRNA | AA124355.1 | - | 3 | 172 | ||
| AY148487.1 | - | 2 | 91 | |||
| BC019553.1 | - | 3 | 163 | 4456658 | ||
| BY096725.1 | - | 3 | 178 | |||
| BY097425.1 | - | 3 | 186 | |||

2210407C18Rik
Genomic Map