siRNA Details
| Gene Symbol: Pcnxl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144868.3 | NP_659117.2 | 11 | 3046 | ||
| GenBank mRNA | AF237953.1 | - | 9 | 1063 | ||
| BC025838.1 | - | 5 | 549 | |||
| BC038056.1 | - | 6 | 1154 | |||
| BC054751.1 | - | 11 | 2325 | |||
| BC055289.1 | - | 12 | 2849 | |||
| BC096538.1 | - | 11 | 3046 | |||

Pcnxl3
Genomic Map